missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)benzaldehyde, 97%, ACROS Organics™

Chemical Identifiers

CAS 454-89-7
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H5F3O
Linear Formula CF3C6H4CHO
Formula Weight 174.12
MDL Number MFCD00003373
Synonym α, α, α-Trifluoro-m-tolualdehyde
Chemical Name or Material 3-(Trifluoromethyl)benzaldehyde, 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC188440050 SDS Thermo Fisher Scientific
188440050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC188440250 SDS Thermo Fisher Scientific
188440250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

454-89-7
100.0
C8H5F3O
174.12
α, α, α-Trifluoro-m-tolualdehyde
≥96.0%
96.0
97%
CF3C6H4CHO
MFCD00003373
3-(Trifluoromethyl)benzaldehyde, 97%

Specifications

1.3g/mL
5g
Authentic
100.0
1.4639 to 1.4659 (20°C, 589nm)
97%
CF3C6H4CHO
MFCD00003373
α, α, α-Trifluoro-m-tolualdehyde
≥96.0%
Clear colorless to very slightly orange
454-89-7
96.0
Glass bottle
Liquid
C8H5F3O
174.12
07, III, 1016
3-(Trifluoromethyl)benzaldehyde, 97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 207-228-1

TSCA : TSCA