Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Trifluoromethyl-alpha-toluenesulfonyl)acetic acid, 98%

$236.93 - $236.93

Chemical Identifiers

Chemical Name or Material (3-Trifluoromethyl-alpha-toluenesulfonyl)acetic acid
CAS 773108-68-2
Molecular Formula C10H9F3O4S
Formula Weight 282.25g/mol
MDL Number MFCD06208211
Synonym 3-(Trifluoromethyl)benzylsulfonylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27811ME

 
alfa aesar™
H27811-ME
500mg Each for $236.93
Description & Specifications

Specifications

Melting Point ≈160°C
Quantity 500mg
Chemical Name or Material (3-Trifluoromethyl-alpha-toluenesulfonyl)acetic acid
CAS 773108-68-2
Assay 98%
Molecular Formula C10H9F3O4S
Formula Weight 282.25g/mol
MDL Number MFCD06208211
Synonym 3-(Trifluoromethyl)benzylsulfonylacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.