Please login to your online account to display your discounted pricing

3'-(Trifluoromethyl)acetophenone, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3'-(Trifluoromethyl)acetophenone
CAS 349-76-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C9H7F3O
Linear Formula CF3C6H4COCH3
Formula Weight 188.15
MDL Number MFCD00000391
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC214440250

  SDS
thermo fisher scientific
AC214440250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC214440050

  SDS
thermo fisher scientific
214440050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (GC)
Physical Form Crystals or Crystalline Powder
Formula Weight 188.15
Quantity 25g
Color White
Density 1.2300g/mL
Fieser 13,226
Beilstein 07, III, 1057
CAS Max % 100.0
Refractive Index 1.4610 to 1.4640 (20°C, 589nm)
Chemical Name or Material 3'-(Trifluoromethyl)acetophenone
Molecular Formula C9H7F3O
MDL Number MFCD00000391
CAS Min % 98.5
Linear Formula CF3C6H4COCH3
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 349-76-8
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.