Please login to your online account to display your discounted pricing

3'-(Trifluoromethyl)acetophenone, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  214440050

 View more versions of this product

Catalog No. AC214440050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Assay ≥98.5% (GC)
Quantity 5g
Color White
Beilstein 07, III, 1057
Fieser 13,226
MDL Number MFCD00000391
Chemical Name or Material 3'-(Trifluoromethyl)acetophenone
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 349-76-8
CAS Min % 98.5
Molecular Formula C9H7F3O
CAS Max % 100.0
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Refractive Index 1.4610 to 1.4640 (20°C, 589nm)
Packaging Glass bottle
Linear Formula CF3C6H4COCH3
Infrared Spectrum Authentic
Formula Weight 188.15
Density 1.2300g/mL

Specifications

Chemical Name or Material 3'-(Trifluoromethyl)acetophenone
CAS 349-76-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C9H7F3O
Linear Formula CF3C6H4COCH3
Formula Weight 188.15
MDL Number MFCD00000391
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents