Please login to your online account to display your discounted pricing

3'-(Trifluoromethyl)acetophenone, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  214440010

 View more versions of this product

Catalog No. AC214440010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Physical Form Crystals or Crystalline Powder
Formula Weight 188.15
Quantity 1g
Color White
Beilstein 07, III, 1057
Fieser 13,226
Density 1.2300g/mL
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4610 to 1.4640 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula CF3C6H4COCH3
Chemical Name or Material 3'-(Trifluoromethyl)acetophenone
Infrared Spectrum Authentic
MDL Number MFCD00000391
CAS 349-76-8
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Molecular Formula C9H7F3O
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3'-(Trifluoromethyl)acetophenone
CAS 349-76-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C9H7F3O
Linear Formula CF3C6H4COCH3
Formula Weight 188.15
MDL Number MFCD00000391
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents