missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-(Trifluoromethyl)acetanilide, 98+%

Manufacturer:  Alfa Aesar™ A1609809

 View more versions of this product

Catalog No. AAA1609809



Description

Description

Specifications

Specifications

3'-(Trifluoromethyl)acetanilide
103°C to 106°C
351-36-0
MFCD00000383
3'-trifluoromethyl acetanilide, 3'-trifluoromethylacetanilide, 3-acetamidobenzotrifluoride, 3-trifluoromethyl acetanilide, 3-trifluoromethylacetanilide, acetanilide, 3-trifluoromethyl, m-acetaminobenzotrifluoride, m-trifluoromethylacetanilide, n-3-trifluoromethyl phenyl acetamide, usaf ma-14
CC(=O)NC1=CC=CC(=C1)C(F)(F)F
203.164
203.16
Odorless
10g
C9H8F3NO
2213222
HNIPNANLYHXYDE-UHFFFAOYSA-N
N-[3-(trifluoromethyl)phenyl]acetamide
9595
≥98%

Chemical Identifiers

351-36-0
203.164
HNIPNANLYHXYDE-UHFFFAOYSA-N
9595
CC(=O)NC1=CC=CC(=C1)C(F)(F)F
C9H8F3NO
MFCD00000383
3'-trifluoromethyl acetanilide, 3'-trifluoromethylacetanilide, 3-acetamidobenzotrifluoride, 3-trifluoromethyl acetanilide, 3-trifluoromethylacetanilide, acetanilide, 3-trifluoromethyl, m-acetaminobenzotrifluoride, m-trifluoromethylacetanilide, n-3-trifluoromethyl phenyl acetamide, usaf ma-14
N-[3-(trifluoromethyl)phenyl]acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 206-512-2

RTECSNumber : AN3740000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications