Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Trifluoromethyl-3-azetidinol hydrochloride, 96%

Chemical Identifiers

Chemical Name or Material 3-Trifluoromethyl-3-azetidinol hydrochloride
CAS 848192-96-1
Formula Weight 177.55
MDL Number MFCD13248564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66227MD

 
alfa aesar™
H66227MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 177.55
Chemical Name or Material 3-Trifluoromethyl-3-azetidinol hydrochloride
MDL Number MFCD13248564
Physical Form Powder
Quantity 250mg
Color White to yellow
Assay 96%
CAS 848192-96-1