missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethy)pyridine-2-carboxylic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC452390010

 View more versions of this product

Catalog No. AC452390010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(Trifluoromethy)pyridine-2-carboxylic acid
1g
96% min. (HPLC)
2-pyridinecarboxylic acid, 3-trifluoromethyl, 3-trifluoromethyl picolinic acid, 3-trifluoromethyl pyridine-2-carboxylic acid, 3-trifluoromethyl-2-picolinic acid, 3-trifluoromethyl-2-pyridinecarboxylic acid, 3-trifluoromethylpicolinic acid, acmc-209qkb, ksc447o0j, trifluoromethyl-2-picolinic acid
C1=CC(=C(N=C1)C(=O)O)C(F)(F)F
191.11
191.11
Glass Bottle
87407-12-3
C7H4F3NO2
DMWLBOPLZYJGPT-UHFFFAOYSA-N
3-(trifluoromethyl)pyridine-2-carboxylic acid
13233844
97%

Chemical Identifiers

87407-12-3
191.11
2-pyridinecarboxylic acid, 3-trifluoromethyl, 3-trifluoromethyl picolinic acid, 3-trifluoromethyl pyridine-2-carboxylic acid, 3-trifluoromethyl-2-picolinic acid, 3-trifluoromethyl-2-pyridinecarboxylic acid, 3-trifluoromethylpicolinic acid, acmc-209qkb, ksc447o0j, trifluoromethyl-2-picolinic acid
3-(trifluoromethyl)pyridine-2-carboxylic acid
C7H4F3NO2
DMWLBOPLZYJGPT-UHFFFAOYSA-N
13233844
C1=CC(=C(N=C1)C(=O)O)C(F)(F)F
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents