Please login to your online account to display your discounted pricing

3-(Trifluoromethy)pyridine-2-carboxylic acid, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethy)pyridine-2-carboxylic acid
Assay Percent Range 97%
Molecular Formula C7H4F3NO2
Formula Weight 191.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC452390010

  SDS
thermo fisher scientific
AC452390010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC452390050

  SDS
thermo fisher scientific
AC452390050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.11
Quantity 1g
CAS 87407-12-3
Molecular Formula C7H4F3NO2
Chemical Name or Material 3-(Trifluoromethy)pyridine-2-carboxylic acid
Assay Percent Range 97%