Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)salicylaldehyde, 99%

$173.28 - $433.95

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)salicylaldehyde
CAS 497959-31-6
Molecular Formula C8H5F3O3
Formula Weight 206.12
MDL Number MFCD03788527
Synonym 2-Hydroxy-3-(trifluoromethoxy)benzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2614703

  SDS
alfa aesar™
H2614703
1g Each for $433.95

AAH26147MD

  SDS
alfa aesar™
H26147MD
250mg Each for $173.28
Description & Specifications

Specifications

Formula Weight 206.12
Chemical Name or Material 3-(Trifluoromethoxy)salicylaldehyde
Assay 99%
Molecular Formula C8H5F3O3
MDL Number MFCD03788527
CAS 497959-31-6
Synonym 2-Hydroxy-3-(trifluoromethoxy)benzaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 58° to 60°C
Quantity 1g

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.