missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethoxy)phenol, TCI America™

Manufacturer:  TCI America T161525G

 View more versions of this product

Catalog No. T161525G



Specifications

Specifications

3-(Trifluoromethoxy)phenol
Yellow
827-99-6
MFCD00040987
UWLJERQTLRORJN-UHFFFAOYSA-N
3-(trifluoromethoxy)phenol
2733261
Clear Liquid at 20°C
80°C
25g
C7H5F3O2
3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, 3-trifluoromethoxy phenol, 3-trifluoromethoxy phenol, pubchem1505, m-trifluoromethoxyphenol, pubchem10391
C1=CC(=CC(=C1)OC(F)(F)F)O
178.11
178.11
≥98.0% (GC)

Chemical Identifiers

827-99-6
178.11
UWLJERQTLRORJN-UHFFFAOYSA-N
2733261
C1=CC(=CC(=C1)OC(F)(F)F)O
C7H5F3O2
MFCD00040987
3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, 3-trifluoromethoxy phenol, 3-trifluoromethoxy phenol, pubchem1505, m-trifluoromethoxyphenol, pubchem10391
3-(trifluoromethoxy)phenol
Safety and Handling

Safety and Handling

TSCA : No

Documents