missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethoxy)phenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 308700010

 View more versions of this product

Catalog No. AC308700010


 

Specifications Description & Specifications

Specifications

3-(Trifluoromethoxy)phenol
Clear colorless to yellow
827-99-6
100.0
Glass bottle
≥97.5%
C7H5F3O2
178.11
1.3700g/mL
1g
97.5
Authentic
Liquid
98%
CF3OC6H4OH
MFCD00040987

Chemical Identifiers

827-99-6
100.0
C7H5F3O2
178.11
3-(Trifluoromethoxy)phenol
97.5
98%
CF3OC6H4OH
MFCD00040987
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning