Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)phenol, 98%

$92.22 - $360.12

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)phenol
CAS 827-99-6
Molecular Formula C7H5F3O2
Formula Weight 178.11
MDL Number MFCD00040987
Beilstein 1868036
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2019603

  SDS
alfa aesar™
B2019603
1g Each for $92.22

AAB2019606

  SDS
alfa aesar™
B2019606
5g Each for $360.12
Description & Specifications

Specifications

Formula Weight 178.11
Beilstein 1868036
Chemical Name or Material 3-(Trifluoromethoxy)phenol
CAS 827-99-6
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00040987
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Boiling Point 68° to 69°C (12mmHg)
Assay 98%
Health Hazard 1 Warning
Flash Point 84°C (183°F)
Quantity 1g
Molecular Formula C7H5F3O2
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.