Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)cinnamic acid, 96%

$86.19 - $86.19

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)cinnamic acid
CAS 168833-80-5
Molecular Formula C10H7F3O3
Formula Weight 232.16
MDL Number MFCD00066337
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2156001

  SDS
alfa aesar™
B2156001
0.25g Each for $86.19
Description & Specifications

Specifications

Formula Weight 232.16
Chemical Name or Material 3-(Trifluoromethoxy)cinnamic acid
Assay 96%
Molecular Formula C10H7F3O3
MDL Number MFCD00066337
CAS 168833-80-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 92° to 95°C
Quantity 0.25g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.