Please login to your online account to display your discounted pricing

3-(trifluoromethoxy)benzylamine, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC375840010

 View more versions of this product

Catalog No. AC375840010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 191.15
Quantity 1g
Color Clear colorless to light yellow
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4510 to 1.4540 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C8H8F3NO
Linear Formula CF3OC6H4CH2NH2
CAS 93071-75-1
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-(Trifluoromethoxy)benzylamine
Infrared Spectrum Authentic
MDL Number MFCD00061267
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 3-(Trifluoromethoxy)benzylamine
CAS 93071-75-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8F3NO
Linear Formula CF3OC6H4CH2NH2
Formula Weight 191.15
MDL Number MFCD00061267
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents