Please login to your online account to display your discounted pricing

3-(trifluoromethoxy)benzylamine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)benzylamine
CAS 93071-75-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H8F3NO
Linear Formula CF3OC6H4CH2NH2
Formula Weight 191.15
MDL Number MFCD00061267
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375840010

  SDS
thermo fisher scientific
AC375840010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375840050

  SDS
thermo fisher scientific
AC375840050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 191.15
Quantity 1g
Color Clear colorless to light yellow
CAS Max % 100.0
Linear Formula CF3OC6H4CH2NH2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H8F3NO
CAS 93071-75-1
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS Min % 97.5
Packaging Glass bottle
Refractive Index 1.4510 to 1.4540 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00061267
Chemical Name or Material 3-(Trifluoromethoxy)benzylamine
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.