Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)benzyl alcohol, 98+%

$105.58 - $429.46

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)benzyl alcohol
CAS 50823-90-0
Molecular Formula C8H7F3O2
Formula Weight 192.14
MDL Number MFCD00061270
Beilstein 2094145
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2012103

 
alfa aesar™
B2012103
1g Each for $105.58

AAB2012106

 
alfa aesar™
B2012106
5g Each for $429.46
Description & Specifications

Specifications

Formula Weight 192.14
Chemical Name or Material 3-(Trifluoromethoxy)benzyl alcohol
Assay ≥98%
Molecular Formula C8H7F3O2
MDL Number MFCD00061270
CAS 50823-90-0
Beilstein 2094145
Boiling Point 97° to 98°C (11mmHg)
Quantity 1g