Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)benzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)benzoyl chloride
CAS 86270-03-3
Molecular Formula C8H4ClF3O2
Formula Weight 224.57
MDL Number MFCD00061268
UN Number UN3265
Beilstein 7006050
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2008403

  SDS
alfa aesar™
B2008403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2008406

  SDS
alfa aesar™
B2008406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.57
Chemical Name or Material 3-(Trifluoromethoxy)benzoyl chloride
Assay 97%
Molecular Formula C8H4ClF3O2
MDL Number MFCD00061268
CAS 86270-03-3
UN Number UN3265
Beilstein 7006050
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g

Applications
3-(Trifluoromethoxy)benzoyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible with oxidizing agents, bases and alcohols.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.