missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Trifluoromethoxy)benzonitrile 98.0+%, TCI America™
$61.38 - $182.49
Chemical Identifiers
CAS | 52771-22-9 |
---|---|
Molecular Formula | C8H4F3NO |
Molecular Weight (g/mol) | 187.12 |
MDL Number | MFCD00040959 |
InChI Key | DCZAPXGEZYVQNX-UHFFFAOYSA-N |
Synonym | 3-trifluoromethoxy benzonitrile, m-trifluoromethoxybenzonitrile, benzonitrile, 3-trifluoromethoxy, 3-trifluoromethoxy-benzonitrile, 3-trifluoromethoxy benzenecarbonitrile, pubchem4825, acmc-1art8, ksc493e1j, m-trifluoromethoxy benzonitrile |
PubChem CID | 142962 |
IUPAC Name | 3-(trifluoromethoxy)benzonitrile |
SMILES | FC(F)(F)OC1=CC=CC(=C1)C#N |
Chemical Identifiers
52771-22-9 | |
187.12 | |
DCZAPXGEZYVQNX-UHFFFAOYSA-N | |
142962 | |
FC(F)(F)OC1=CC=CC(=C1)C#N |
C8H4F3NO | |
MFCD00040959 | |
3-trifluoromethoxy benzonitrile, m-trifluoromethoxybenzonitrile, benzonitrile, 3-trifluoromethoxy, 3-trifluoromethoxy-benzonitrile, 3-trifluoromethoxy benzenecarbonitrile, pubchem4825, acmc-1art8, ksc493e1j, m-trifluoromethoxy benzonitrile | |
3-(trifluoromethoxy)benzonitrile |
Specifications
52771-22-9 | |
C8H4F3NO | |
1 g | |
3-trifluoromethoxy benzonitrile, m-trifluoromethoxybenzonitrile, benzonitrile, 3-trifluoromethoxy, 3-trifluoromethoxy-benzonitrile, 3-trifluoromethoxy benzenecarbonitrile, pubchem4825, acmc-1art8, ksc493e1j, m-trifluoromethoxy benzonitrile | |
FC(F)(F)OC1=CC=CC(=C1)C#N | |
187.12 | |
187.12 | |
Liquid |
Yellow | |
MFCD00040959 | |
3276 | |
DCZAPXGEZYVQNX-UHFFFAOYSA-N | |
3-(trifluoromethoxy)benzonitrile | |
142962 | |
≥98.0% (GC) | |
3-(Trifluoromethoxy)benzonitrile |
Safety and Handling
TSCA : No