Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)benzoic acid, 99%

$51.88 - $781.96

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)benzoic acid
CAS 1014-81-9
Molecular Formula C8H5F3O3
Formula Weight 206.12
MDL Number MFCD00041501
Beilstein 2579574
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1810603

 
alfa aesar™
A1810603
1g Each for $51.88

AAA1810606

 
alfa aesar™
A1810606
5g Each for $187.80

AAA1810614

 
alfa aesar™
A1810614
25g Each for $781.96
Description & Specifications

Specifications

Formula Weight 206.12
Chemical Name or Material 3-(Trifluoromethoxy)benzoic acid
Molecular Formula C8H5F3O3
MDL Number MFCD00041501
CAS 1014-81-9
Beilstein 2579574
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 88° to 89°C
Quantity 1g
Assay 99%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.