Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)benzamide, 99%

$65.24 - $65.24

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)benzamide
CAS 658-91-3
Molecular Formula C8H6F3NO2
Formula Weight 205.14
MDL Number MFCD00041512
Beilstein 2645012
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1219503

  SDS
alfa aesar™
L1219503
1g Each for $65.24
Description & Specifications

Specifications

Formula Weight 205.14
Chemical Name or Material 3-(Trifluoromethoxy)benzamide
Assay 99%
MDL Number MFCD00041512
CAS 658-91-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 115° to 117°C
Molecular Formula C8H6F3NO2
Beilstein 2645012
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.