missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)benzamide, 99%

Manufacturer:  Alfa Aesar™ L1219503

 View more versions of this product

Catalog No. AAL1219503



Specifications

Specifications

3-(Trifluoromethoxy)benzamide
1g
C8H6F3NO2
2645012
RRANUIMYSXUNCN-UHFFFAOYSA-N
3-(trifluoromethoxy)benzamide
522189
99%
115°C to 117°C
658-91-3
MFCD00041512
3-trifluoromethoxy benzamide, 3-trifluoromethoxy-benzamide, 3-trifluoromethyloxy benzamide, acmc-1b717, benzamide,3-trifluoromethoxy, 3-trifluoromethoxy-benzamide, benzamide, 3-trifluoromethoxy
C1=CC(=CC(=C1)OC(F)(F)F)C(=O)N
205.136
205.14

Chemical Identifiers

658-91-3
205.136
RRANUIMYSXUNCN-UHFFFAOYSA-N
522189
C1=CC(=CC(=C1)OC(F)(F)F)C(=O)N
C8H6F3NO2
MFCD00041512
3-trifluoromethoxy benzamide, 3-trifluoromethoxy-benzamide, 3-trifluoromethyloxy benzamide, acmc-1b717, benzamide,3-trifluoromethoxy, 3-trifluoromethoxy-benzamide, benzamide, 3-trifluoromethoxy
3-(trifluoromethoxy)benzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents