Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethoxy)benzaldehyde, 97%

$323.96 - $323.96

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)benzaldehyde
CAS 52771-21-8
Molecular Formula C8H5F3O2
Formula Weight 190.12
MDL Number MFCD00042406
Beilstein 7267831
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1929609

  SDS
alfa aesar™
A1929609
10g Each for $323.96
Description & Specifications

Specifications

Formula Weight 190.12
Chemical Name or Material 3-(Trifluoromethoxy)benzaldehyde
Assay 97%
Molecular Formula C8H5F3O2
MDL Number MFCD00042406
CAS 52771-21-8
Beilstein 7267831
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 72°C (161°F)
Boiling Point 83° to 86°C (24mmHg)
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.