missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethoxy)benzaldehyde, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 297650050

 View more versions of this product

Catalog No. AC297650050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
190.12
Clear yellow
94.0
Glass bottle
Authentic
95%
52771-21-8
C8H5F3O2
Liquid
5g
1.33g/mL
100.0
1.4530 to 1.4550 (20°C, 589nm)
3-(Trifluoromethoxy)benzaldehyde
CF3OC6H4CHO
95%
MFCD00042406

Chemical Identifiers

52771-21-8
100.0
C8H5F3O2
190.12
3-(Trifluoromethoxy)benzaldehyde, 95%
94.0
95%
CF3OC6H4CHO
MFCD00042406
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA