missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethoxy)benzaldehyde, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC297650010

 View more versions of this product

Catalog No. AC297650010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
Liquid
Clear yellow
100.0
Glass bottle
1.4530 to 1.4550 (20°C, 589nm)
C8H5F3O2
CF3OC6H4CHO
MFCD00042406
190.12
1g
1.33g/mL
94.0
Authentic
95%
3-(Trifluoromethoxy)benzaldehyde
95%
52771-21-8

Chemical Identifiers

52771-21-8
100.0
C8H5F3O2
190.12
3-(Trifluoromethoxy)benzaldehyde, 95%
94.0
95%
CF3OC6H4CHO
MFCD00042406
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA