Please login to your online account to display your discounted pricing

3-(Trifluoromethoxy)benzaldehyde, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)benzaldehyde
Name Note 95%
CAS 52771-21-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H5F3O2
Linear Formula CF3OC6H4CHO
Formula Weight 190.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC297650010

  SDS
thermo fisher scientific
AC297650010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC297650050

  SDS
thermo fisher scientific
297650050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 190.12
Quantity 1g
Color Clear yellow
TSCA TSCA
Density 1.33g/mL
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Chemical Name or Material 3-(Trifluoromethoxy)benzaldehyde
Name Note 95%
Assay Percent Range 95%
Molecular Formula C8H5F3O2
Linear Formula CF3OC6H4CHO
MDL Number MFCD00042406
CAS 52771-21-8
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.