Please login to your online account to display your discounted pricing

3-(Trifluoromethoxy)aniline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethoxy)aniline
Name Note 97%
CAS 1535-73-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H6F3NO
Linear Formula CF3OC6H4NH2
Formula Weight 177.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC297635000

  SDS
thermo fisher scientific
297635000
Glass Bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 177.12
Quantity 500mg
Color Clear yellow to orange
Packaging Glass Bottle
EINECS Number 216-256-3
TSCA TSCA
Density 1.32g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-(Trifluoromethoxy)aniline
Infrared Spectrum Authentic
Refractive Index 1.4650 to 1.4670 (20°C, 589nm)
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7H6F3NO
Linear Formula CF3OC6H4NH2
MDL Number MFCD00041511
CAS 1535-73-5
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.