Learn More
3-(Trifluoroacetyl)pyridine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H66214MD
Description
3-(Trifluoroacetyl)pyridine is used as medical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Trifluoroacetyl)pyridine is used as medical intermediates.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
3-(Trifluoroacetyl)pyridine | |
C7H6F3NO | |
250 mg | |
2,2,2-trifluoro-1-pyridin-3-yl ethanol, 2,2,2-trifluoro-1-pyridin-3-yl ethan-1-ol, 2,2,2-trifluoro-1-3-pyridyl ethanol, alpha-trifluoromethyl pyridine-3-methanol, 10.14272/lhxkphqpuhhfqw-uhfffaoysa-n, 3-2,2,2-trifluoro-1-hydroxyethyl pyridine, doi:10.14272/lhxkphqpuhhfqw-uhfffaoysa-n | |
C1=CC(=CN=C1)C(C(F)(F)F)O | |
177.126 | |
175.11 | |
Solid |
33284-21-8 | |
MFCD16067922 | |
4800857 | |
LHXKPHQPUHHFQW-UHFFFAOYSA-N | |
2,2,2-trifluoro-1-pyridin-3-ylethanol | |
11217557 | |
95% |
Chemical Identifiers
33284-21-8 | |
177.126 | |
LHXKPHQPUHHFQW-UHFFFAOYSA-N | |
11217557 | |
C1=CC(=CN=C1)C(C(F)(F)F)O |
C7H6F3NO | |
MFCD16067922 | |
2,2,2-trifluoro-1-pyridin-3-yl ethanol, 2,2,2-trifluoro-1-pyridin-3-yl ethan-1-ol, 2,2,2-trifluoro-1-3-pyridyl ethanol, alpha-trifluoromethyl pyridine-3-methanol, 10.14272/lhxkphqpuhhfqw-uhfffaoysa-n, 3-2,2,2-trifluoro-1-hydroxyethyl pyridine, doi:10.14272/lhxkphqpuhhfqw-uhfffaoysa-n | |
2,2,2-trifluoro-1-pyridin-3-ylethanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only