missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoroacetyl)indole, 97%

$51.04 - $234.41

Chemical Identifiers

CAS 14618-45-2
Molecular Formula C10H6F3NO
Molecular Weight (g/mol) 213.159
MDL Number MFCD00182114
InChI Key LCMDCXWSHDFQKP-UHFFFAOYSA-N
Synonym 3-trifluoroacetyl indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone, 3-trifluoroacetylindole, indole3trifluoroacetyl, 2,2,2-trifluoro-1-1h-indol-3-yl-ethanone, 3-trifluoroacetyl-1h-indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one, ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl, 2,2,2-trifluoro-1-indol-3-ylethan-1-one
PubChem CID 589126
IUPAC Name 2,2,2-trifluoro-1-(1H-indol-3-yl)ethanone
SMILES C1=CC=C2C(=C1)C(=CN2)C(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6600003 View Documents Alfa Aesar™
H6600003
1g Each for $51.04
 
AAH6600006 View Documents Alfa Aesar™
H6600006
5g Each for $234.41
 
Specifications

Chemical Identifiers

14618-45-2
213.159
LCMDCXWSHDFQKP-UHFFFAOYSA-N
589126
C1=CC=C2C(=C1)C(=CN2)C(=O)C(F)(F)F
C10H6F3NO
MFCD00182114
3-trifluoroacetyl indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone, 3-trifluoroacetylindole, indole3trifluoroacetyl, 2,2,2-trifluoro-1-1h-indol-3-yl-ethanone, 3-trifluoroacetyl-1h-indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one, ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl, 2,2,2-trifluoro-1-indol-3-ylethan-1-one
2,2,2-trifluoro-1-(1H-indol-3-yl)ethanone

Specifications

211°C to 214°C
14618-45-2
MFCD00182114
3-trifluoroacetyl indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl-1-ethanone, 3-trifluoroacetylindole, indole3trifluoroacetyl, 2,2,2-trifluoro-1-1h-indol-3-yl-ethanone, 3-trifluoroacetyl-1h-indole, 2,2,2-trifluoro-1-1h-indol-3-yl ethan-1-one, ethanone, 2,2,2-trifluoro-1-1h-indol-3-yl, 2,2,2-trifluoro-1-indol-3-ylethan-1-one
C1=CC=C2C(=C1)C(=CN2)C(=O)C(F)(F)F
213.159
213.16
97%
1g
C10H6F3NO
181739
LCMDCXWSHDFQKP-UHFFFAOYSA-N
2,2,2-trifluoro-1-(1H-indol-3-yl)ethanone
589126
Solid
3-(Trifluoroacetyl)indole