missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Tri-n-butylstannyl)pyridine, 97%, Thermo Scientific™

 

Manufacturer:  Thermo Fisher Scientific H5126803

Catalog No. AAH5126803



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-(Tri-n-butylstannyl)pyridine
1.153
MFCD00052054
3-1,1,1-tributylstannyl pyridine, 3-pyridyltri-n-butyltin, 3-pyridyltributylstannane, 3-pyridyltributyltin, 3-tri-n-butylstannyl pyridine, 3-tributylstannanyl-pyridine, 3-tributylstannyl pyridine, 3-tributylstannylpyridine, 3-tris but-1-ylstannyl pyridine, pyridine, 3-tributylstannyl
CFQJBWKKHCMCGJ-UHFFFAOYSA-N
tributyl(pyridin-3-yl)stannane
3483848
97%
59020-10-9
C17H31NSn
UN2788
1g
CCCC[Sn](CCCC)(CCCC)C1=CN=CC=C1
368.152
368.15

Chemical Identifiers

59020-10-9
368.152
CFQJBWKKHCMCGJ-UHFFFAOYSA-N
3483848
CCCC[Sn](CCCC)(CCCC)C1=CN=CC=C1
C17H31NSn
MFCD00052054
3-1,1,1-tributylstannyl pyridine, 3-pyridyltri-n-butyltin, 3-pyridyltributylstannane, 3-pyridyltributyltin, 3-tri-n-butylstannyl pyridine, 3-tributylstannanyl-pyridine, 3-tributylstannyl pyridine, 3-tributylstannylpyridine, 3-tris but-1-ylstannyl pyridine, pyridine, 3-tributylstannyl
tributyl(pyridin-3-yl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P260-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H360FD-H372

DOTInformation : Hazard Class: 6.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only