Alfa Aesar™ 3-(trans-2-Cyanovinyl)benzeneboronic acid, 95%
Manufacturer:  Alfa Aesar™ H53270MD
 View more versions of this product
Catalog No. AAH53270MD
Specifications
Specifications
3-(trans-2-Cyanovinyl)benzeneboronic acid
850568-53-5
MFCD04973741
YDSLGHLJEJPWNC-DUXPYHPUSA-N
[3-[(E)-2-cyanoethenyl]phenyl]boronic acid
6294465
95%
250mg
C9H8BNO2
e-3-2-cyanovinyl phenyl boronic acid, 3-e-2-cyanovinyl phenylboronic acid, 3-3-boronophenyl acrylonitrile, 2e-3-3-boronophenyl acrylonitrile, 3-e-cyanoethenyl benzeneboronic acid, 3-e-2-cyanovinylbenzeneboronic acid, 3-1e-2-cyanoeth-1-en-1-yl phenylboronic acid, 3-2-cyanoeth-1-en-1-yl phenylboronic acid, 3-boronoacrylonitrile, 3-e-2-cyanoethenyl phenyl boronic acid
B(C1=CC(=CC=C1)C=CC#N)(O)O
172.978
172.98
Chemical Identifiers
850568-53-5
172.978
YDSLGHLJEJPWNC-DUXPYHPUSA-N
6294465
B(C1=CC(=CC=C1)C=CC#N)(O)O
C9H8BNO2
MFCD04973741
e-3-2-cyanovinyl phenyl boronic acid, 3-e-2-cyanovinyl phenylboronic acid, 3-3-boronophenyl acrylonitrile, 2e-3-3-boronophenyl acrylonitrile, 3-e-cyanoethenyl benzeneboronic acid, 3-e-2-cyanovinylbenzeneboronic acid, 3-1e-2-cyanoeth-1-en-1-yl phenylboronic acid, 3-2-cyanoeth-1-en-1-yl phenylboronic acid, 3-boronoacrylonitrile, 3-e-2-cyanoethenyl phenyl boronic acid
[3-[(E)-2-cyanoethenyl]phenyl]boronic acid
Safety and Handling
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
SDS Documents