Please login to your online account to display your discounted pricing

3-Toluenesulfonyl chloride, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC364630010

 View more versions of this product

Catalog No. AC364630010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (exCl)
Formula Weight 190.64
Physical Form Liquid
Quantity 1g
Color Clear yellow
UN Number 3265
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
CAS 1899-93-0
Molecular Formula C7H7ClO2S
Chemical Name or Material 3-Toluenesulfonyl chloride
Assay Percent Range 97%
MDL Number MFCD00051747
Infrared Spectrum Authentic
Refractive Index 1.5485 to 1.5495 (20°C, 589nm)
Linear Formula CH3C6H4SOCl
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Toluenesulfonyl chloride
CAS 1899-93-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (exCl)
Assay Percent Range 97%
Molecular Formula C7H7ClO2S
Linear Formula CH3C6H4SOCl
Formula Weight 190.64
MDL Number MFCD00051747
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents