missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Thiophenesulfonyl chloride, ≥95%, Maybridge™

Manufacturer:  Fisher Scientific Company CC13103CB

 View more versions of this product

Catalog No. CC13103CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Thiophenesulfonyl chloride
51175-71-4
100.0
C4H3ClO2S2
3-chlorosulphonyl thiophene, 3-thienyl sulfonyl chloride, 3-thienyl-sulfonyl chloride, 3-thienylsulfonyl chloride, 3-thiophene sulfonyl chloride, 3-thiophenesulfonyl chloride, 3-thiophenesulfonylchloride, 3-thiophenesulphonylchloride, acmc-209ksa, chloro-3-thienylsulfone
ClS(=O)(=O)C1=CSC=C1
182.64
182.65
250mg
96.5
97%
MFCD02682011
YSPWSQNKRBSICH-UHFFFAOYSA-N
thiophene-3-sulfonyl chloride
2776378
97%

Chemical Identifiers

51175-71-4
182.64
YSPWSQNKRBSICH-UHFFFAOYSA-N
2776378
ClS(=O)(=O)C1=CSC=C1
C4H3ClO2S2
MFCD02682011
3-chlorosulphonyl thiophene, 3-thienyl sulfonyl chloride, 3-thienyl-sulfonyl chloride, 3-thienylsulfonyl chloride, 3-thiophene sulfonyl chloride, 3-thiophenesulfonyl chloride, 3-thiophenesulfonylchloride, 3-thiophenesulphonylchloride, acmc-209ksa, chloro-3-thienylsulfone
thiophene-3-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents
Product Certifications