Learn More
3-Thiophenemethylamine, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6148703
Description
3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Thiophenemethylamineis an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store in cool, dry place in tightly closed container. With good ventilation. Store away from air.
Specifications
3-Thiophenemethylamine | |
1.13 | |
C5H7NS | |
MFCD01529872 | |
UN2735 | |
3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine | |
DUDAKCCDHRNMDJ-UHFFFAOYSA-N | |
thiophen-3-ylmethanamine | |
2776381 | |
97% |
27757-86-4 | |
102°C (15 mmHg) | |
1.566 | |
1 g | |
Air sensitive | |
Slightly soluble in water. | |
C1=CSC=C1CN | |
113.178 | |
113.18 |
Chemical Identifiers
27757-86-4 | |
113.178 | |
DUDAKCCDHRNMDJ-UHFFFAOYSA-N | |
2776381 | |
C1=CSC=C1CN |
C5H7NS | |
MFCD01529872 | |
3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine | |
thiophen-3-ylmethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only