missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Thiophenemethanol, TCI America™

Manufacturer:  TCI America T166025G

 View more versions of this product

Catalog No. T166025G



Specifications

Specifications

3-Thiophenemethanol
Yellow
71637-34-8
MFCD00014534
BOWIFWCBNWWZOG-UHFFFAOYSA-N
thiophen-3-ylmethanol
123570
Clear Liquid at 20°C
73°C
25g
C5H6OS
3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
C1=CSC=C1CO
114.162
114.16
≥96.0% (GC)

Chemical Identifiers

71637-34-8
114.162
BOWIFWCBNWWZOG-UHFFFAOYSA-N
123570
C1=CSC=C1CO
C5H6OS
MFCD00014534
3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
thiophen-3-ylmethanol
Safety and Handling

Safety and Handling

TSCA : No

Documents