Learn More
3-Thiophenemethanol, 97%
CAS: 71637-34-8 | C5H6OS | 114.16 g/mol
Supplier: Thermo Scientific Chemicals L0193506
Description
3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 3-Thiophenemethanol | |
| 1.21 | |
| 110°C (230°F) | |
| 1.565 | |
| 5 g | |
| 3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol | |
| BOWIFWCBNWWZOG-UHFFFAOYSA-N | |
| thiophen-3-ylmethanol | |
| 123570 | |
| 98% |
| 71637-34-8 | |
| 86°C to 88°C (10 mmHg) | |
| C5H6OS | |
| MFCD00014534 | |
| 106460 | |
| Slightly soluble in water. | |
| OCC1=CSC=C1 | |
| 114.16 | |
| 114.17 |
Chemical Identifiers
| 71637-34-8 | |
| 114.16 | |
| BOWIFWCBNWWZOG-UHFFFAOYSA-N | |
| 123570 |
| C5H6OS | |
| MFCD00014534 | |
| 3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol | |
| OCC1=CSC=C1 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 275-741-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only