Learn More
3-Thiopheneethanol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1513806
Description
3-Thiopheneethanol is used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Thiopheneethanol is used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene
Solubility
Sparingly soluble in water(0.059 g/L at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications
3-Thiopheneethanol | |
1.159 | |
92°C (197°F) | |
1.553 | |
5 g | |
2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415 | |
YYPNNBPPDFTQFX-UHFFFAOYSA-N | |
2-thiophen-3-ylethanol | |
83731 | |
98% |
13781-67-4 | |
110°C to 111°C (14 mmHg) | |
C6H8OS | |
MFCD00009766 | |
107080 | |
Sparingly soluble in water(0.059g/L at 25°C). | |
OCCC1=CSC=C1 | |
128.19 | |
128.19 |
Chemical Identifiers
13781-67-4 | |
128.19 | |
YYPNNBPPDFTQFX-UHFFFAOYSA-N | |
83731 | |
OCCC1=CSC=C1 |
C6H8OS | |
MFCD00009766 | |
2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415 | |
2-thiophen-3-ylethanol |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P233-P240-P241-P242-P243-P280-P370+P378q-P374-P501c
H227
EINECSNumber : 237-434-7
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only