Learn More
3-Thiopheneacetic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205370500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Thiopheneacetic acid | |
6964-21-2 | |
100.0 | |
White to Beige | |
98% | |
C6H6O2S | |
50 g | |
3-thiopheneacetic acid, 3-thienylacetic acid, 2-thiophen-3-yl acetic acid, thiophene-3-acetic acid, thiophen-3-yl-acetic acid, 3-thiophenacetic acid, thien-3-ylacetic acid, 3-thiopheneethanoic acid, thiophen-3-ylacetic acid, 2-3-thienyl acetic acid | |
C1=CSC=C1CC(=O)O | |
142.17 | |
142.17 | |
Crystalline Flakes or Powder |
98% | |
97.5 | |
77°C to 80°C | |
Authentic | |
Glass bottle | |
MFCD00005473 | |
18, IV, 4066 | |
RCNOGGGBSSVMAS-UHFFFAOYSA-N | |
2-thiophen-3-ylacetic acid | |
23404 | |
98% |
Chemical Identifiers
6964-21-2 | |
142.17 | |
RCNOGGGBSSVMAS-UHFFFAOYSA-N | |
23404 | |
C1=CSC=C1CC(=O)O |
C6H6O2S | |
MFCD00005473 | |
3-thiopheneacetic acid, 3-thienylacetic acid, 2-thiophen-3-yl acetic acid, thiophene-3-acetic acid, thiophen-3-yl-acetic acid, 3-thiophenacetic acid, thien-3-ylacetic acid, 3-thiopheneethanoic acid, thiophen-3-ylacetic acid, 2-3-thienyl acetic acid | |
2-thiophen-3-ylacetic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 230-166-1
RTECSNumber : XM7570000
TSCA : TSCA
RUO – Research Use Only