Learn More
3-Thiopheneacetic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1070022
Description
3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications
3-Thiopheneacetic acid | |
77°C to 81°C | |
C6H6O2S | |
100 g | |
113622 | |
Very soluble in water. | |
C1=CSC=C1CC(=O)O | |
142.172 | |
142.18 |
6964-21-2 | |
129°C (2.5 mmHg) | |
MFCD00005473 | |
UN3261 | |
3-thiopheneacetic acid, 3-thienylacetic acid, 2-thiophen-3-yl acetic acid, thiophene-3-acetic acid, thiophen-3-yl-acetic acid, 3-thiophenacetic acid, thien-3-ylacetic acid, 3-thiopheneethanoic acid, thiophen-3-ylacetic acid, 2-3-thienyl acetic acid | |
RCNOGGGBSSVMAS-UHFFFAOYSA-N | |
2-thiophen-3-ylacetic acid | |
23404 | |
98% |
Chemical Identifiers
6964-21-2 | |
142.172 | |
RCNOGGGBSSVMAS-UHFFFAOYSA-N | |
23404 | |
C1=CSC=C1CC(=O)O |
C6H6O2S | |
MFCD00005473 | |
3-thiopheneacetic acid, 3-thienylacetic acid, 2-thiophen-3-yl acetic acid, thiophene-3-acetic acid, thiophen-3-yl-acetic acid, 3-thiophenacetic acid, thien-3-ylacetic acid, 3-thiopheneethanoic acid, thiophen-3-ylacetic acid, 2-3-thienyl acetic acid | |
2-thiophen-3-ylacetic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 230-166-1
RTECSNumber : XM7570000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only