Please login to your online account to display your discounted pricing

3-Thienylboronic acid, 97%

$14.65 - $14.65

Chemical Identifiers

Chemical Name or Material 3-Thienylboronic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H5BO2S
Formula Weight 127.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC13112DA

  SDS
fisher scientific company
CC13112DA
Amber glass bottle 1g Each for $14.65
Description & Specifications

Specifications

Formula Weight 127.96
Quantity 1g
Assay 97%
CAS 6165-69-1
Assay Percent Range 97%
Molecular Formula C4H5BO2S
Chemical Name or Material 3-Thienylboronic acid
Packaging Amber glass bottle