Please login to your online account to display your discounted pricing

3-Thienylboronic acid, 97%


Quantity 5g

Manufacturer: fisher scientific company  CC13112DE

 View more versions of this product

Catalog No. CC13112DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 5g
Chemical Name or Material 3-Thienylboronic acid
CAS 6165-69-1
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H5BO2S
Formula Weight 127.96g/mol

Specifications

Chemical Name or Material 3-Thienylboronic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H5BO2S
Formula Weight 127.96g/mol