Learn More
(3-Thien-2-ylphenyl)methanol, ≥97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC44509DE
Specifications
(3-Thien-2-ylphenyl)methanol | |
96.5 | |
97% | |
MFCD06203108 | |
3-thien-2-ylphenyl methanol, 3-2-thienyl benzyl alcohol, 3-thiophen-2-yl phenyl methanol, 3-2-thienyl phenyl methanol, benzenemethanol,3-2-thienyl, acmc-1c9kv, 3-thiophen-2-ylphenyl methanol, 3-thiophen-2-yl benzyl alcohol, 3-2-thienyl phenyl methan-1-ol | |
OCC1=CC=CC(=C1)C1=CC=CS1 | |
190.26 | |
190.27 |
103669-00-7 | |
100.0 | |
C11H10OS | |
5g | |
SATFADRDDGOLJP-UHFFFAOYSA-N | |
(3-thiophen-2-ylphenyl)methanol | |
7537586 | |
97% |
Chemical Identifiers
103669-00-7 | |
190.26 | |
SATFADRDDGOLJP-UHFFFAOYSA-N | |
7537586 | |
OCC1=CC=CC(=C1)C1=CC=CS1 |
C11H10OS | |
MFCD06203108 | |
3-thien-2-ylphenyl methanol, 3-2-thienyl benzyl alcohol, 3-thiophen-2-yl phenyl methanol, 3-2-thienyl phenyl methanol, benzenemethanol,3-2-thienyl, acmc-1c9kv, 3-thiophen-2-ylphenyl methanol, 3-thiophen-2-yl benzyl alcohol, 3-2-thienyl phenyl methan-1-ol | |
(3-thiophen-2-ylphenyl)methanol |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning