missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Thien-2-ylbenzaldehyde, ≥97%, Maybridge

Chemical Identifiers

CAS 103668-99-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H8OS
Formula Weight 188.25
MDL Number MFCD04038019
Chemical Name or Material 3-Thien-2-ylbenzaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC44504CB SDS Fisher Scientific Company
CC44504CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

103668-99-1
100.0
C11H8OS
MFCD04038019
97%
96.5
97%
188.25
3-Thien-2-ylbenzaldehyde

Specifications

188.25
100.0
97%
Amber Glass Bottle
MFCD04038019
97%
250mg
C11H8OS
96.5
103668-99-1
3-Thien-2-ylbenzaldehyde

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.