missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Tetrazol-5-yl)phenylboronic acid, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445560050

 View more versions of this product

Catalog No. AC445560050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(Tetrazol-5-yl)phenylboronic acid
775351-30-9
C7H7BN4O2
XMJKBDSKRITXBW-UHFFFAOYSA-N
[3-(2H-tetrazol-5-yl)phenyl]boronic acid
46737996
96%
5g
95% min. (HPLC)
3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-1h-tetrazol-5-yl phenylboronic acid, 3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-2h-tetrazol-5-yl phenyl boronic acid, 3-2h-tetrazol-5-yl phenyl-boronic acid, 3-2h-tetrazol-5-yl-phenyl-boronic acid, 3-tetrazol-5-yl phenylboronic acid, acmc-209pal, boronic acid,b-3-2h-tetrazol-5-yl phenyl
B(C1=CC(=CC=C1)C2=NNN=N2)(O)O
189.97
189.97

Chemical Identifiers

775351-30-9
189.97
3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-1h-tetrazol-5-yl phenylboronic acid, 3-2h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 3-2h-tetrazol-5-yl phenyl boronic acid, 3-2h-tetrazol-5-yl phenyl-boronic acid, 3-2h-tetrazol-5-yl-phenyl-boronic acid, 3-tetrazol-5-yl phenylboronic acid, acmc-209pal, boronic acid,b-3-2h-tetrazol-5-yl phenyl
[3-(2H-tetrazol-5-yl)phenyl]boronic acid
C7H7BN4O2
XMJKBDSKRITXBW-UHFFFAOYSA-N
46737996
B(C1=CC(=CC=C1)C2=NNN=N2)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

Documents