missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester, 90%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445540050
Specifications
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester | |
5g | |
88% min. (GC) | |
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl oxy oxane, 4,4,5,5-tetramethyl-2-3-oxan-2-yloxy prop-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-3-tetrahydro-2h-pyran-2-yl oxy prop-1-en-1-yl-1,3,2-dioxaborolane | |
B1(OC(C(O1)(C)C)(C)C)C=CCOC2CCCCO2 | |
268.16 | |
268.16 |
79°C (0.1 Torr) | |
642066-70-4 | |
C14H25BO4 | |
MHSOBXCZCRNELG-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-[3-(oxan-2-yloxy)prop-1-enyl]-1,3,2-dioxaborolane | |
53406456 | |
90% |
Chemical Identifiers
642066-70-4 | |
268.16 | |
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl oxy oxane, 4,4,5,5-tetramethyl-2-3-oxan-2-yloxy prop-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-3-tetrahydro-2h-pyran-2-yl oxy prop-1-en-1-yl-1,3,2-dioxaborolane | |
4,4,5,5-tetramethyl-2-[3-(oxan-2-yloxy)prop-1-enyl]-1,3,2-dioxaborolane |
C14H25BO4 | |
MHSOBXCZCRNELG-UHFFFAOYSA-N | |
53406456 | |
B1(OC(C(O1)(C)C)(C)C)C=CCOC2CCCCO2 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning