Learn More
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 445540010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester | |
79°C (0.1 Torr) | |
Glass Bottle | |
1g | |
MHSOBXCZCRNELG-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-[3-(oxan-2-yloxy)prop-1-enyl]-1,3,2-dioxaborolane | |
53406456 | |
90% |
642066-70-4 | |
88% min. (GC) | |
C14H25BO4 | |
4,4,5,5-tetramethyl-2-3-tetrahydro-2h-pyran-2-yl oxy prop-1-en-1-yl-1,3,2-dioxaborolane, 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl oxy oxane, 4,4,5,5-tetramethyl-2-3-oxan-2-yloxy prop-1-en-1-yl-1,3,2-dioxaborolane | |
B1(OC(C(O1)(C)C)(C)C)C=CCOC2CCCCO2 | |
268.16 | |
268.16 |
Chemical Identifiers
642066-70-4 | |
268.16 | |
4,4,5,5-tetramethyl-2-3-tetrahydro-2h-pyran-2-yl oxy prop-1-en-1-yl-1,3,2-dioxaborolane, 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl oxy oxane, 4,4,5,5-tetramethyl-2-3-oxan-2-yloxy prop-1-en-1-yl-1,3,2-dioxaborolane | |
4,4,5,5-tetramethyl-2-[3-(oxan-2-yloxy)prop-1-enyl]-1,3,2-dioxaborolane |
C14H25BO4 | |
MHSOBXCZCRNELG-UHFFFAOYSA-N | |
53406456 | |
B1(OC(C(O1)(C)C)(C)C)C=CCOC2CCCCO2 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO â Research Use Only