Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Tetrahydrofurfurylaminocarbonyl)benzeneboronic acid pinacol ester, 97%

$137.24 - $137.24

Chemical Identifiers

Chemical Name or Material 3-(Tetrahydrofurfurylaminocarbonyl)benzeneboronic acid pinacol ester
CAS 1073355-06-2
Molecular Formula C18H26BNO4
Formula Weight 331.21g/mol
MDL Number MFCD09266189
Synonym 3-(Tetrahydrofurfurylaminocarbonyl)phenylboronic acid pinacol ester; N-Tetrahydrofurfuryl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2813006

 
alfa aesar™
H28130-06
5g Each for $137.24
Description & Specifications

Specifications

Quantity 5g
Melting Point 92° to 94°C
Quantity 5g
Chemical Name or Material 3-(Tetrahydrofurfurylaminocarbonyl)benzeneboronic acid pinacol ester
CAS 1073355-06-2
Assay 97%
Molecular Formula C18H26BNO4
Formula Weight 331.21g/mol
MDL Number MFCD09266189
Synonym 3-(Tetrahydrofurfurylaminocarbonyl)phenylboronic acid pinacol ester; N-Tetrahydrofurfuryl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide