missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Tetradecylthiophene, TCI America™

Manufacturer:  TCI America T310425G

 View more versions of this product

Catalog No. T310425G



Specifications

Specifications

3-Tetradecylthiophene
25g
C18H32S
CAEIOINMYGTXNS-UHFFFAOYSA-N
3-tetradecylthiophene
14900816
Clear Liquid at 20°C
Yellow
110851-66-6
MFCD09750675
CCCCCCCCCCCCCCC1=CSC=C1
280.514
280.51
≥98.0% (GC)

Chemical Identifiers

110851-66-6
280.514
CAEIOINMYGTXNS-UHFFFAOYSA-N
3-tetradecylthiophene
C18H32S
MFCD09750675
14900816
CCCCCCCCCCCCCCC1=CSC=C1
Safety and Handling

Safety and Handling

TSCA : No

Documents