Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-tert-Butyltoluene, 99%

Chemical Identifiers

Chemical Name or Material 3-tert-Butyltoluene
CAS 1075-38-3
Molecular Formula C11H16
Formula Weight 148.25g/mol
MDL Number MFCD00059210
UN Number UN2667
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3266803

 
alfa aesar™
H32668-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3266806

 
alfa aesar™
H32668-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 189°C
Melting Point −41°C
Odor Aromatic
Quantity 1g
Chemical Name or Material 3-tert-Butyltoluene
CAS 1075-38-3
Assay 99%
Molecular Formula C11H16
Formula Weight 148.25g/mol
MDL Number MFCD00059210
UN Number UN2667
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H227-H302-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BUTYLTOLUENES
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.