missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(tert-Butyldimethylsilyloxy)phenylboronic acid, +96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398340010

 View more versions of this product

Catalog No. AC398340010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Powder
Off-white
96.0
3-(tert-Butyldimethylsilyloxy)phenylboronic acid
261621-12-9
MFCD03701505
(CH3)CSi(CH3)2OC6H4B(OH)2
252.19
1g
100.0
Authentic
96+%
C12H21BO3Si
Glass bottle

Chemical Identifiers

261621-12-9
100.0
C12H21BO3Si
252.19
3-(tert-Butyldimethylsilyloxy)phenylboronic acid
96.0
96+%
(CH3)CSi(CH3)2OC6H4B(OH)2
MFCD03701505
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.